Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000563403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000563403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000563403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000575674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000575674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000575674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
ENST00000575674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012960.hg.1 |
ENST00000261381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012960.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012960.hg.1 |
TC16000904.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012961.hg.1 |
ENST00000261381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012961.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012961.hg.1 |
TC16000904.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600173180.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173180.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600173180.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173180.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600173180.hg.1 |
TC1600009580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600173179.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173179.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600173179.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173179.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600173179.hg.1 |
TC1600009580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000261381.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000563403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000563403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000563403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000568226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000575674.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000575674.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000575674.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
ENST00000575674.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009580.hg.1 |
NM_022166.3 |
Refseq |