Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005906.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005906.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005906.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005906.hg.1 |
TC14000474.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005908.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005908.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005908.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005908.hg.1 |
TC14000474.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005909.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005909.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005909.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005909.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005909.hg.1 |
TC14000474.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400154524.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154524.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154524.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154524.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154524.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154524.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154524.hg.1 |
TC1400007712.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154526.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154526.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154526.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154526.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154526.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154526.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154526.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154526.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154526.hg.1 |
TC1400007712.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007712.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154525.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154525.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154525.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154525.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154525.hg.1 |
ENST00000555504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154525.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154525.hg.1 |
TC1400007712.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154522.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154522.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154522.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154522.hg.1 |
ENST00000286639.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154522.hg.1 |
ENST00000555795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154522.hg.1 |
NM_006399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154522.hg.1 |
TC1400007712.hg.1 |
Clariom D TC |