Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023957.hg.1 |
TC07001580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023957.hg.1 |
ENST00000341119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023957.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000483810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000483810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000483810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023955.hg.1 |
TC07001580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023955.hg.1 |
ENST00000341119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023955.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000444363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000483810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000483810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000483810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000491499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011710.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000470328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183661.hg.1 |
TC0700011710.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000483810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000341119.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000429674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000446236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000483810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
ENST00000496348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
NM_018843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183659.hg.1 |
TC0700011710.hg.1 |
Clariom D TC |