Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005123.hg.1 |
ENST00000256433.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005123.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005123.hg.1 |
TC18000493.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005121.hg.1 |
ENST00000256433.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005121.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005121.hg.1 |
TC18000493.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800164135.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800164135.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800164135.hg.1 |
TC1800009281.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800164135.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800164135.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009281.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800164134.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800164134.hg.1 |
NM_016097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800164134.hg.1 |
TC1800009281.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800164134.hg.1 |
ENST00000256433.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800164134.hg.1 |
NM_016097.4 |
Refseq |