Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004622.hg.1 |
ENST00000400752.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004622.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004622.hg.1 |
TC16000326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004625.hg.1 |
ENST00000400752.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004625.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004625.hg.1 |
TC16000326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004623.hg.1 |
ENST00000400752.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004623.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004623.hg.1 |
TC16000326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
ENST00000572964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153620.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153620.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153620.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153620.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153620.hg.1 |
TC1600007424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
ENST00000572964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007424.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153621.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153621.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153621.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153621.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153621.hg.1 |
TC1600007424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600153624.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153624.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153624.hg.1 |
ENST00000400752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153624.hg.1 |
NM_152338.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153624.hg.1 |
TC1600007424.hg.1 |
Clariom D TC |