Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128920.2 | NP_001122392.2 | 10 - 11 | 1660 | 156 | |
XM_005267642.4 | XP_005267699.2 | 10 - 11 | 1660 | 156 | ||
XM_017021291.1 | XP_016876780.1 | 10 - 11 | 1660 | 156 | ||
XM_017021292.1 | XP_016876781.1 | 10 - 11 | 1660 | 156 | ||
XM_017021293.1 | XP_016876782.1 | 11 - 12 | 1135 | 156 | ||
XM_017021297.1 | XP_016876786.1 | 9 - 10 | 1417 | 156 | ||
XM_017021299.1 | XP_016876788.1 | 9 - 10 | 1468 | 156 | ||
XM_017021305.1 | XP_016876794.1 | 8 - 9 | 894 | 156 | ||
XM_017021308.1 | XP_016876797.1 | 9 - 10 | 1611 | 156 | ||
GenBank mRNA | AF170723.1 | - | 1 - 2 | 97 | 156 | |
AK296643.1 | - | 3 - 4 | 297 | 156 | ||
M80359.1 | - | 10 - 11 | 1165 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Homo sapiens Probe spans exons |