Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027482.hg.1 |
TC02001708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027482.hg.1 |
ENST00000389048.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027482.hg.1 |
NM_004304.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027481.hg.1 |
TC02001708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027481.hg.1 |
ENST00000389048.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027481.hg.1 |
NM_004304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188581.hg.1 |
ENST00000389048.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188581.hg.1 |
ENST00000431873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188581.hg.1 |
ENST00000618119.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188581.hg.1 |
NM_004304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188581.hg.1 |
ENST00000389048.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188581.hg.1 |
ENST00000431873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188581.hg.1 |
ENST00000618119.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188581.hg.1 |
NM_004304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188581.hg.1 |
TC0200012140.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
ENST00000389048.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
ENST00000431873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
ENST00000453137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
ENST00000618119.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
NM_004304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
ENST00000389048.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
ENST00000431873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
ENST00000453137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
ENST00000618119.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
NM_004304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188580.hg.1 |
TC0200012140.hg.1 |
Clariom D TC |