Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010897.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010897.hg.1 |
ENST00000270594.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010897.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010897.hg.1 |
TC19000768.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008603.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167174.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167174.hg.1 |
NM_033068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167174.hg.1 |
TC1900008603.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167174.hg.1 |
ENST00000270593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167174.hg.1 |
NM_033068.2 |
Refseq |