Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
TC1900008627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167407.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
TC1900008627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167408.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008627.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
TC1900008627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
ENST00000614626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
ENST00000619985.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167409.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
NR_047531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
TC1900008627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
ENST00000593736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
ENST00000611992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
ENST00000618545.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
NR_047529.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
NR_047530.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167405.hg.1 |
NR_047531.2 |
Refseq |