Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000261366.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000395354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000460265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000463908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000463908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000463908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000463908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000463908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000472034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000472034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000472034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000472034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000472034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000472034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000472034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000472034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000492190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000492190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000492190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000492190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000492190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000492190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000492190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000494185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000494185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000494185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000494185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000494185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
ENST00000504788.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_001198557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008544.hg.1 |
NM_005573.3 |
Refseq |