Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017934.5 | NP_060404.3 | 12 - 13 | 1367 | 121 | |
XM_005248729.4 | XP_005248786.1 | 12 - 13 | 1367 | 121 | ||
XM_011535918.2 | XP_011534220.1 | 9 - 10 | 1023 | 121 | ||
XM_011535919.1 | XP_011534221.1 | 12 - 13 | 1367 | 121 | ||
XM_017010989.1 | XP_016866478.1 | 13 - 14 | 1445 | 121 | ||
XM_017010990.1 | XP_016866479.1 | 10 - 11 | 1043 | 121 | ||
GenBank mRNA | AJ303102.1 | - | 12 - 13 | 1154 | 121 | |
AK057039.1 | - | 12 - 13 | 1309 | 121 | ||
BC021905.1 | - | 12 - 13 | 1256 | 121 | ||
BC137488.1 | - | 12 - 13 | 1247 | 9021488 | 121 | |
DQ924532.1 | - | 12 - 13 | 1141 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |