Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017934.5 | NP_060404.3 | 29 - 30 | 3607 | 88 | |
XM_005248729.4 | XP_005248786.1 | 29 - 30 | 3604 | 88 | ||
XM_011535918.2 | XP_011534220.1 | 26 - 27 | 3263 | 88 | ||
XM_017010989.1 | XP_016866478.1 | 31 - 32 | 3749 | 88 | ||
XM_017010990.1 | XP_016866479.1 | 28 - 29 | 3347 | 88 | ||
GenBank mRNA | AF310250.1 | - | 6 - 7 | 614 | 88 | |
AJ303102.1 | - | 29 - 30 | 3394 | 88 | ||
AK000712.1 | - | 2 - 3 | 237 | 88 | ||
AL161957.1 | - | 7 - 8 | 670 | 88 | ||
BC008909.2 | - | 4 - 5 | 335 | 88 | ||
BC064611.1 | - | 1 - 2 | 76 | 88 | ||
BC081569.1 | - | 7 - 8 | 650 | 88 | ||
BC137488.1 | - | 29 - 30 | 3487 | 9021488 | 88 | |
DQ924532.1 | - | 29 - 30 | 3381 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |