Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023213.hg.1 |
ENST00000323777.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023213.hg.1 |
ENST00000412540.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023213.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023213.hg.1 |
TC17001733.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700190566.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190566.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190566.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190566.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190566.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190566.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190566.hg.1 |
TC1700011257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700190567.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190567.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190567.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190567.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190567.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190567.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190567.hg.1 |
TC1700011257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000323777.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
ENST00000617946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011257.hg.1 |
NM_001080439.2 |
Refseq |