Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023670.hg.1 |
TC01001534.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023670.hg.1 |
ENST00000367662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023670.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023671.hg.1 |
TC01001534.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023671.hg.1 |
ENST00000367662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023671.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000479836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000479836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000479836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000479836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000486075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000486075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000486075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000493665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
ENST00000493665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178706.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178706.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178706.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178706.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178706.hg.1 |
TC0100010713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000479836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000479836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000479836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000479836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000486075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000486075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000486075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000493665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
ENST00000493665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010713.hg.1 |
NM_021936.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178707.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178707.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178707.hg.1 |
ENST00000367662.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178707.hg.1 |
NM_020318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178707.hg.1 |
TC0100010713.hg.1 |
Clariom D TC |