Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
ENST00000323541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
ENST00000323578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
ENST00000336845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
ENST00000429789.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
ENST00000436108.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
ENST00000449825.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
ENST00000454838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
ENST00000528581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
NM_001142674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
NM_001142675.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
NM_001142676.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
NM_001142677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
NM_023947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015843.hg.1 |
TC11001253.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000323578.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000429789.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000436108.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000449825.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000454838.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000524832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000528521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000528581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000531010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000531859.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000534254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_001142674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_001142675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_001142676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_001142677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_023947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000323578.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000429789.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000436108.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000449825.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000454838.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000524832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000528521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000528581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000531010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000531859.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
ENST00000534254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_001142674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_001142675.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_001142676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_001142677.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
NM_023947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173340.hg.1 |
TC1100009709.hg.1 |
Clariom D TC |