Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
ENST00000258187.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
ENST00000356752.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
ENST00000394279.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
ENST00000394282.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
ENST00000421602.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
NM_001130487.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
NM_001242833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
NM_001242834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
NM_001242835.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
NM_001242836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
NM_020465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
NM_022910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
NR_040072.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007077.hg.1 |
TC16000503.hg.1 |
HTA TC |