Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003133.hg.1 |
TC6_dbb_hap3000161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003133.hg.1 |
NM_021177.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003119.hg.1 |
TC6_qbl_hap6000162.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003119.hg.1 |
NM_021177.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017924.hg.1 |
TC06001545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017924.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017924.hg.1 |
ENST00000491421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017924.hg.1 |
NM_021177.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001284.hg.1 |
TC6_apd_hap1000086.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001284.hg.1 |
NM_021177.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003226.hg.1 |
TC6_cox_hap2000172.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003226.hg.1 |
NM_021177.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002893.hg.1 |
TC6_mcf_hap5000149.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002893.hg.1 |
NM_021177.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000475835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000475835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000475835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000475835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000475835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000475835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000475835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000475835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000477182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011457.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000375661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000470083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000470086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000491421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
ENST00000493387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177700.hg.1 |
TC0600011457.hg.1 |
Clariom D TC |