Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001992.1 | NP_001001992.1 | 16 - 17 | 2399 | 150 | |
NM_001032410.1 | NP_001027582.1 | 17 - 18 | 2465 | 150 | ||
NM_006447.2 | NP_006438.1 | 16 - 17 | 2402 | 150 | ||
XM_017028257.1 | XP_016883746.1 | 17 - 18 | 2549 | 150 | ||
XM_017028258.1 | XP_016883747.1 | 17 - 18 | 2428 | 150 | ||
XM_017028259.1 | XP_016883748.1 | 17 - 18 | 2400 | 150 | ||
XM_017028260.1 | XP_016883749.1 | 15 - 16 | 2145 | 150 | ||
XM_017028261.1 | XP_016883750.1 | 17 - 18 | 2392 | 150 | ||
GenBank mRNA | AF113219.1 | - | 12 - 13 | 1787 | 150 | |
AF126736.1 | - | 16 - 17 | 2328 | 150 | ||
AK023247.1 | - | 11 - 12 | 1751 | 150 | ||
AK025104.1 | - | 16 - 17 | 2365 | 150 | ||
AK222681.1 | - | 16 - 17 | 2331 | 150 | ||
AK222884.1 | - | 16 - 17 | 2324 | 150 | ||
AK294284.1 | - | 15 - 16 | 2263 | 150 | ||
AK300771.1 | - | 7 - 8 | 1135 | 150 | ||
AK302247.1 | - | 10 - 11 | 1511 | 150 | ||
AY333928.1 | - | 15 - 16 | 3222 | 150 | ||
BC030777.2 | - | 16 - 17 | 2355 | 4824914 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |