Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8019333 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019332 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8019331 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
242378_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11735286_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11735286_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026819.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026819.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026819.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026819.hg.1 |
TC17001967.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026822.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026822.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026822.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026822.hg.1 |
TC17001967.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026824.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026824.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026824.hg.1 |
TC17001967.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026820.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026820.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026820.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026820.hg.1 |
TC17001967.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026821.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026821.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026821.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026821.hg.1 |
TC17001967.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700208384.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208384.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208384.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208384.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208384.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208384.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208384.hg.1 |
TC1700012485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700208386.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208386.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208386.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208386.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208386.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208386.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208386.hg.1 |
TC1700012485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700208385.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208385.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208385.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208385.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208385.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208385.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208385.hg.1 |
TC1700012485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700208387.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208387.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208387.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208387.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208387.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208387.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208387.hg.1 |
TC1700012485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
ENST00000330655.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012485.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208389.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208389.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208389.hg.1 |
ENST00000409745.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208389.hg.1 |
NM_001145113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208389.hg.1 |
TC1700012485.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
242378_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
242378_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242378_at4 |
- |
- |
Human Genome U95 Set |
71202_at |
- |
- |
Human Genome U95 Set |
82164_PM_at |
- |
- |
Human Genome U95 Set |
82164_at |
- |
- |
Human Genome U95 Set |
71202_PM_at |
- |
- |
Human Genome U133 Set |
242378_at |
- |
- |
Human Genome U133 Set |
242378_PM_at |
- |
- |