Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002813.hg.1 |
ENST00000291634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002813.hg.1 |
ENST00000397826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002813.hg.1 |
ENST00000479127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002813.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002813.hg.1 |
TC21000240.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002811.hg.1 |
ENST00000291634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002811.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002811.hg.1 |
TC21000240.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000485207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000485207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000485207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
ENST00000485207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002814.hg.1 |
ENST00000291634.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002814.hg.1 |
ENST00000397826.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002814.hg.1 |
ENST00000479127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002814.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002814.hg.1 |
TC21000240.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000485207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000485207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000485207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
ENST00000485207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007399.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150060.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150060.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150060.hg.1 |
TC2100007399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100150060.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150060.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
NM_058190.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
TC2100007399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
ENST00000291634.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
ENST00000397826.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
ENST00000458015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
ENST00000479127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150062.hg.1 |
NM_058190.3 |
Refseq |