Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007135.hg.1 |
ENST00000355481.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007135.hg.1 |
ENST00000359596.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007135.hg.1 |
ENST00000360985.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007135.hg.1 |
NM_000540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007135.hg.1 |
NM_001042723.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007135.hg.1 |
TC19000530.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
ENST00000355481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
ENST00000359596.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
ENST00000360985.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
ENST00000594335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
NM_000540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
NM_001042723.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
TC1900008017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
ENST00000355481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
ENST00000359596.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
ENST00000360985.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
ENST00000594335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
NM_000540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900160173.hg.1 |
NM_001042723.1 |
Refseq |