Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013196.hg.1 |
TC09001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013196.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013196.hg.1 |
ENST00000536631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013196.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
ENST00000410237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167349.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167349.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167349.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167349.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167349.hg.1 |
TC0900009844.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000397149.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
ENST00000410237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009844.hg.1 |
NM_018225.2 |
Refseq |