Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001214.hg.1 |
TC06000115.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001214.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001214.hg.1 |
ENST00000542476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001214.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009625.hg.1 |
ENST00000427218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001211.hg.1 |
TC06000115.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001211.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001211.hg.1 |
ENST00000542476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001211.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001212.hg.1 |
TC06000115.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001212.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001212.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001213.hg.1 |
TC06000115.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001213.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001213.hg.1 |
ENST00000542476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001213.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001208.hg.1 |
TC06000115.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001208.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006623.hg.1 |
ENST00000435433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010853.hg.1 |
ENST00000528813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148373.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148373.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148373.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148373.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148373.hg.1 |
TC0600007092.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600148374.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148374.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148374.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148374.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148374.hg.1 |
TC0600007092.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007092.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148372.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148372.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148372.hg.1 |
ENST00000259963.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148372.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148372.hg.1 |
TC0600007092.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600148370.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148370.hg.1 |
NM_016255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148370.hg.1 |
TC0600007092.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00006623.hg.1 |
ENST00000435433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009625.hg.1 |
ENST00000427218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010853.hg.1 |
ENST00000528813.1 |
Ensembl |