Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000303795.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000445692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
ENST00000529851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016464.hg.1 |
NM_173545.2 |
Refseq |