Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002714.hg.1 |
TC02000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002714.hg.1 |
ENST00000407426.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002714.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002715.hg.1 |
TC02000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002715.hg.1 |
ENST00000407426.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002715.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000434238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000434238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000577887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
ENST00000579969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007111.hg.1 |
NR_002741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002718.hg.1 |
TC02000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002718.hg.1 |
ENST00000407426.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002718.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148999.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148999.hg.1 |
ENST00000434238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148999.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148999.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148999.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148999.hg.1 |
ENST00000434238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148999.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148999.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148999.hg.1 |
TC0200007111.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000296126.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000434238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000434238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000440983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000446643.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000466067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000577887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
ENST00000579969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007111.hg.1 |
NR_002741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148995.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148995.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148995.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148995.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148995.hg.1 |
TC0200007111.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200148996.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148996.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148996.hg.1 |
ENST00000407426.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148996.hg.1 |
NM_015131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148996.hg.1 |
TC0200007111.hg.1 |
Clariom D TC |