Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015530.hg.1 |
TC0X001143.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015530.hg.1 |
ENST00000339490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015530.hg.1 |
ENST00000373539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015530.hg.1 |
ENST00000373542.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015530.hg.1 |
ENST00000373545.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015530.hg.1 |
ENST00000541944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015530.hg.1 |
NM_001122670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015530.hg.1 |
NM_001172436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015530.hg.1 |
NM_002637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000339490.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000373539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000373542.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000373545.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000541944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
NM_001122670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
NM_001172436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
NM_002637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000339490.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000373539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000373542.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000373545.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
ENST00000541944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
NM_001122670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
NM_001172436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
NM_002637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168528.hg.1 |
TC0X00010038.hg.1 |
Clariom D TC |