Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018565.hg.1 |
TC01001202.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018565.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018565.hg.1 |
ENST00000368917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018565.hg.1 |
ENST00000368918.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018565.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018564.hg.1 |
TC01001202.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018564.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018564.hg.1 |
ENST00000368917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018564.hg.1 |
ENST00000368918.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018564.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018564.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018566.hg.1 |
TC01001202.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018566.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018566.hg.1 |
ENST00000368917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018566.hg.1 |
ENST00000368918.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018566.hg.1 |
ENST00000446567.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01018566.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171927.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171927.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171927.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171927.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171927.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171927.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171927.hg.1 |
TC0100009939.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100171925.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171925.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171925.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171925.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171925.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171925.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171925.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100171925.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100171925.hg.1 |
TC0100009939.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000368918.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000462520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000467551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000486076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
ENST00000489549.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009939.hg.1 |
NM_144618.2 |
Refseq |