Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006956.hg.1 |
TC06000596.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006956.hg.1 |
ENST00000265354.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006956.hg.1 |
ENST00000457278.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006956.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156271.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156271.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156271.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156271.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156271.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156271.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156271.hg.1 |
TC0600008073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
ENST00000265354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_001292001.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008073.hg.1 |
NM_003131.3 |
Refseq |