Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024023.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024023.hg.1 |
TC17001798.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000561739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000561739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000561739.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000566616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000566616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000566616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000569530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000569530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000569530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192445.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192445.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192445.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192445.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192445.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192445.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192445.hg.1 |
TC1700011442.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700192441.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192441.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192441.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192441.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192441.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192441.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192441.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700192441.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700192441.hg.1 |
TC1700011442.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000561739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000561739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000561739.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000563924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000564866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000566422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000566616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000566616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000566616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000568702.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000569530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000569530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000569530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
ENST00000569967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011442.hg.1 |
NM_000442.4 |
Refseq |