Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000337682.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000448582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000454658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000536836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000536836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000536836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000539409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000542983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000542983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000542983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000542983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000542983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000542983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000542983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000543615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000543615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000543615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000543615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000543615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000543615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000544921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000544921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000544921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000544921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000544921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000544921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000544921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000544921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000622889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
ENST00000622889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135811.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_001135812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010281.hg.1 |
NM_021238.2 |
Refseq |