Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314017.1 | NP_001300946.1 | 4 - 5 | 353 | 109 | |
NM_020939.1 | NP_065990.1 | 4 - 5 | 353 | 109 | ||
XM_005249247.2 | XP_005249304.1 | 4 - 5 | 353 | 109 | ||
XM_011514768.1 | XP_011513070.1 | 4 - 5 | 353 | 109 | ||
XM_011514769.1 | XP_011513071.1 | 4 - 5 | 353 | 109 | ||
XM_011514770.1 | XP_011513072.1 | 5 - 6 | 368 | 109 | ||
XM_011514771.2 | XP_011513073.1 | 4 - 5 | 693 | 109 | ||
XM_011514772.1 | XP_011513074.1 | 4 - 5 | 353 | 109 | ||
XM_017011139.1 | XP_016866628.1 | 2 - 3 | 223 | 109 | ||
GenBank mRNA | AA912142.1 | - | 3 - 4 | 109 | ||
AB046819.1 | - | 4 - 5 | 911 | 109 | ||
AI951562.1 | - | 3 - 4 | 109 | |||
DA265730.1 | - | 4 - 5 | 353 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |