Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040249.hg.1 |
TC02002538.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040249.hg.1 |
ENST00000342016.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040249.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000464393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000464393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000464393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000464393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209339.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209339.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209339.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209339.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209339.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209339.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209339.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209339.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209339.hg.1 |
TC0200014990.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000342016.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000377973.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000461454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000464393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000464393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000464393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000464393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
ENST00000472169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014990.hg.1 |
NM_004882.3 |
Refseq |