Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011394.hg.1 |
ENST00000215793.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011394.hg.1 |
ENST00000439242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011394.hg.1 |
ENST00000536049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011394.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011394.hg.1 |
TC22000650.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000471037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000471037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000471037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000471342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000471342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000471342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000496189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000496189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000496189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000496189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011395.hg.1 |
ENST00000215793.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011395.hg.1 |
ENST00000439242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011395.hg.1 |
ENST00000536049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011395.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011395.hg.1 |
TC22000650.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000411423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000444440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000447376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000463818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000471037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000471037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000471037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000471342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000471342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000471342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000485618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000496189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000496189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000496189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000496189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
ENST00000498259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008434.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160519.hg.1 |
TC2200008434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160519.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160519.hg.1 |
NM_005877.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160519.hg.1 |
ENST00000215793.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160519.hg.1 |
NM_005877.5 |
Refseq |