Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK057612.1 | - | 1 | 968 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |