Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002985.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002985.hg.1 |
TC14000280.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002987.hg.1 |
ENST00000298316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002987.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002987.hg.1 |
TC14000280.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002988.hg.1 |
ENST00000298316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002988.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002988.hg.1 |
TC14000280.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400149241.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149241.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149241.hg.1 |
TC1400007098.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400149244.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149244.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149244.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149244.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149244.hg.1 |
TC1400007098.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400149243.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149243.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149243.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149243.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149243.hg.1 |
TC1400007098.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
ENST00000298316.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007098.hg.1 |
NM_001663.3 |
Refseq |