Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281729.1 | NP_001268658.1 | 3 - 4 | 536 | 68 | |
GenBank mRNA | BM561321.1 | - | 3 - 4 | 506 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |