Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009042.hg.1 |
ENST00000202816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009042.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009042.hg.1 |
TC20000637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009038.hg.1 |
ENST00000202816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009038.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009038.hg.1 |
TC20000637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009039.hg.1 |
ENST00000202816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009039.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009039.hg.1 |
TC20000637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009041.hg.1 |
ENST00000202816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009041.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009041.hg.1 |
TC20000637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009037.hg.1 |
ENST00000202816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009037.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009037.hg.1 |
TC20000637.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000159323.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159323.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159323.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159323.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159323.hg.1 |
TC2000008439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159323.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159323.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159323.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159323.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008439.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159326.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159326.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159326.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159326.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159326.hg.1 |
TC2000008439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159326.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159326.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159326.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159326.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159327.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159327.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159327.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159327.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159327.hg.1 |
TC2000008439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159327.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159327.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159327.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159327.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159324.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159324.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159324.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159324.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159324.hg.1 |
TC2000008439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159324.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159324.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159324.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159324.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159325.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159325.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159325.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159325.hg.1 |
NM_016649.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159325.hg.1 |
TC2000008439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159325.hg.1 |
ENST00000202816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159325.hg.1 |
ENST00000617257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159325.hg.1 |
NM_001276380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159325.hg.1 |
NM_016649.3 |
Refseq |