Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008100.hg.1 |
TC20001744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008100.hg.1 |
ENST00000303103.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008100.hg.1 |
ENST00000339610.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008100.hg.1 |
ENST00000381342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008100.hg.1 |
ENST00000438552.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008100.hg.1 |
ENST00000474384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008100.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008100.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008101.hg.1 |
TC20001744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008101.hg.1 |
ENST00000303103.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008101.hg.1 |
ENST00000339610.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008101.hg.1 |
ENST00000381342.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008101.hg.1 |
ENST00000438552.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008101.hg.1 |
ENST00000474384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008101.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008101.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
TC2000009972.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170945.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
TC2000009972.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170944.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000339610.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000381342.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000438552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
ENST00000474384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_003091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009972.hg.1 |
NM_198216.1 |
Refseq |