Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
ENST00000585099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009114.hg.1 |
NR_039816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010141.hg.1 |
TC09000782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010141.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010143.hg.1 |
TC09000782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010143.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010143.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010147.hg.1 |
TC09000782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010147.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010147.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010147.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010142.hg.1 |
TC09000782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010142.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162074.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162074.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162074.hg.1 |
TC0900009114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162083.hg.1 |
TC0900009114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000356384.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000484822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
ENST00000585099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_001291921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009114.hg.1 |
NR_039816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162075.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162075.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162075.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162075.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162075.hg.1 |
TC0900009114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900162073.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162073.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162073.hg.1 |
TC0900009114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900162076.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162076.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162076.hg.1 |
ENST00000481739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162076.hg.1 |
NM_002957.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162076.hg.1 |
TC0900009114.hg.1 |
Clariom D TC |