Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025180.3 | NP_079456.2 | 12 - 13 | 1827 | 123 | |
XM_005247795.4 | XP_005247852.1 | 11 - 12 | 1500 | 123 | ||
XM_005247797.3 | XP_005247854.1 | 11 - 12 | 1755 | 123 | ||
XM_005247799.3 | XP_005247856.1 | 11 - 12 | 1568 | 123 | ||
XM_006713760.3 | XP_006713823.1 | 11 - 12 | 1432 | 123 | ||
XM_011513194.2 | XP_011511496.1 | 11 - 12 | 1549 | 123 | ||
XM_011513197.2 | XP_011511499.1 | 10 - 11 | 1383 | 123 | ||
XM_017007244.1 | XP_016862733.1 | 11 - 12 | 1543 | 123 | ||
XM_017007245.1 | XP_016862734.1 | 10 - 11 | 1402 | 123 | ||
XM_017007246.1 | XP_016862735.1 | 10 - 11 | 1294 | 123 | ||
XM_017007247.1 | XP_016862736.1 | 10 - 11 | 1411 | 123 | ||
XM_017007248.1 | XP_016862737.1 | 10 - 11 | 1578 | 123 | ||
XM_017007249.1 | XP_016862738.1 | 11 - 12 | 1667 | 123 | ||
XM_017007250.1 | XP_016862739.1 | 11 - 12 | 1541 | 123 | ||
XM_017007251.1 | XP_016862740.1 | 11 - 12 | 1539 | 123 | ||
XM_017007252.1 | XP_016862741.1 | 11 - 12 | 1474 | 123 | ||
GenBank mRNA | AK056465.1 | - | 12 - 13 | 1805 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |