Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020859.3 | NP_065910.3 | 9 - 10 | 6303 | 61 | |
XM_005263163.4 | XP_005263220.1 | 8 - 9 | 10757 | 61 | ||
XM_005263164.4 | XP_005263221.1 | 8 - 9 | 5315 | 61 | ||
XM_005263165.4 | XP_005263222.1 | 8 - 9 | 6741 | 61 | ||
XM_006714282.3 | XP_006714345.1 | 9 - 10 | 5458 | 61 | ||
XM_011532157.2 | XP_011530459.2 | 8 - 9 | 6145 | 61 | ||
XM_011532158.2 | XP_011530460.1 | 10 - 11 | 6104 | 61 | ||
XM_011532159.1 | XP_011530461.1 | 9 - 10 | 6293 | 61 | ||
XM_011532160.1 | XP_011530462.1 | 7 - 8 | 5305 | 61 | ||
XM_011532161.2 | XP_011530463.1 | 8 - 9 | 5483 | 61 | ||
XM_017008488.1 | XP_016863977.1 | 9 - 10 | 5461 | 61 | ||
XM_017008489.1 | XP_016863978.1 | 9 - 10 | 5362 | 61 | ||
GenBank mRNA | AB040914.1 | - | 5 - 6 | 3502 | 61 | |
AB055660.1 | - | 9 - 10 | 6304 | 61 | ||
AK128677.1 | - | 5 - 6 | 4805 | 61 | ||
BC007291.1 | - | 5 - 6 | 1664 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons |