Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000471171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000471171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000471171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000471171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000490568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000490568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000490568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
ENST00000490568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044477.hg.1 |
TC01002850.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044477.hg.1 |
ENST00000343435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044477.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044477.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044471.hg.1 |
TC01002850.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044471.hg.1 |
ENST00000343435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044471.hg.1 |
ENST00000370459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044471.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044471.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044473.hg.1 |
TC01002850.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044473.hg.1 |
ENST00000343435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044473.hg.1 |
ENST00000370459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044473.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044476.hg.1 |
TC01002850.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044476.hg.1 |
ENST00000343435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044476.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044476.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210726.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210726.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210726.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210726.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210726.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210726.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210726.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210726.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210726.hg.1 |
TC0100014857.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000471171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000471171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000471171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000471171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000481145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000490568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000490568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000490568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
ENST00000490568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014857.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210732.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210732.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210732.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210732.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210732.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210732.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210732.hg.1 |
TC0100014857.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100210729.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210729.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210729.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210729.hg.1 |
ENST00000370459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210729.hg.1 |
ENST00000443807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210729.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210729.hg.1 |
TC0100014857.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100210733.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210733.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210733.hg.1 |
NM_001134486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210733.hg.1 |
NM_052942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210733.hg.1 |
TC0100014857.hg.1 |
Clariom D TC |