Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024690.2 | NP_078966.2 | 50 - 51 | 40347 | 82 | |
XM_017027486.1 | XP_016882975.1 | 44 - 45 | 39427 | 82 | ||
XM_017027487.1 | XP_016882976.1 | 43 - 44 | 39361 | 82 | ||
XM_017027488.1 | XP_016882977.1 | 44 - 45 | 39427 | 82 | ||
XM_017027489.1 | XP_016882978.1 | 41 - 42 | 39273 | 82 | ||
XM_017027490.1 | XP_016882979.1 | 39 - 40 | 38962 | 82 | ||
XM_017027491.1 | XP_016882980.1 | 44 - 45 | 39427 | 82 | ||
XM_017027493.1 | XP_016882982.1 | 39 - 40 | 38959 | 82 | ||
XM_017027499.1 | XP_016882988.1 | 44 - 45 | 30214 | 82 | ||
XM_017027500.1 | XP_016882989.1 | 43 - 44 | 30133 | 82 | ||
XM_017027501.1 | XP_016882990.1 | 23 - 24 | 2141 | 82 | ||
GenBank mRNA | AF361486.3 | - | 17607 | 82 | ||
AF414442.2 | - | 45975 | 82 | |||
AK024365.1 | - | 2 - 3 | 90 | 82 | ||
AK128856.1 | - | 7 - 8 | 767 | 82 | ||
AK128857.1 | - | 2043 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |