Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000425186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000425186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000425186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000425186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000443887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000443887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
ENST00000443887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003782.hg.1 |
ENST00000339105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003782.hg.1 |
ENST00000376354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003782.hg.1 |
ENST00000376355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003782.hg.1 |
ENST00000376360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003782.hg.1 |
ENST00000444838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003782.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003782.hg.1 |
TC13000358.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000443887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
ENST00000443887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152812.hg.1 |
TC1300007845.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000339105.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000376355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000416504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000419700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000425186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000425186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000425186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000425186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000443887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000443887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
ENST00000443887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NM_206808.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007845.hg.1 |
NR_104592.1 |
Refseq |