Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002879.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002879.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002879.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002877.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002877.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002877.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002876.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002876.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002876.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002878.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002878.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002878.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002870.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002870.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002870.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002873.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002873.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002873.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002882.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002882.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002882.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002880.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002880.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002880.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002874.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002874.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002874.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002881.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002881.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002881.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002872.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002872.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002872.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002871.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002871.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002871.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002875.hg.1 |
ENST00000382675.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002875.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002875.hg.1 |
TC18000247.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800152429.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152429.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152429.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152429.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152429.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152436.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152436.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152436.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152436.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152436.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152431.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152431.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152431.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152431.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152431.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152437.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152437.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152437.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152437.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152437.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152439.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152439.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152439.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152439.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152439.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152435.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152435.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152435.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152435.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152435.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152433.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152433.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152433.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152433.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152433.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152434.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152434.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152434.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152434.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152434.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152438.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152438.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152438.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152438.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152438.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152441.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152441.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152441.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152441.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152441.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152428.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152428.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152428.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152428.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152428.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152430.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152430.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152430.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152430.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152430.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152440.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152440.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152440.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152440.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152440.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007673.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152432.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152432.hg.1 |
NM_001174123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152432.hg.1 |
TC1800007673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152432.hg.1 |
ENST00000579455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152432.hg.1 |
NM_001174123.1 |
Refseq |