Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047507.hg.1 |
TC01003051.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047507.hg.1 |
ENST00000256585.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047507.hg.1 |
ENST00000354219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047507.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047507.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215555.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215555.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215555.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215555.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215555.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215555.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215555.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215555.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215555.hg.1 |
TC0100015477.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000256585.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000354219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000369401.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
ENST00000530654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_001159353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015477.hg.1 |
NM_032044.3 |
Refseq |