Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012325.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012325.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012325.hg.1 |
TC11000988.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100166582.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166582.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166582.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166582.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166582.hg.1 |
TC1100009004.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009004.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166581.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166581.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166581.hg.1 |
ENST00000260270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166581.hg.1 |
NM_004109.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166581.hg.1 |
TC1100009004.hg.1 |
Clariom D TC |