Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035492.hg.1 |
TC01002318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035492.hg.1 |
ENST00000375031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035492.hg.1 |
ENST00000444387.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035492.hg.1 |
ENST00000460804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035492.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035492.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197629.hg.1 |
TC0100013192.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000375031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000444387.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000447746.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000460804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000517430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000518294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
ENST00000519434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103160.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013192.hg.1 |
NM_001103161.1 |
Refseq |