Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000418498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000438351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000438351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000438351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000438351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000438351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000438351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000438351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000438351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000439991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000439991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000439991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000439991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000439991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000439991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000439991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000470595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000470595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000470595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000470595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000481301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000481301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000481301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000546910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
ENST00000551012.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_001301167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012849.hg.1 |
NM_013252.2 |
Refseq |