Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010894.hg.1 |
TC07000695.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010894.hg.1 |
ENST00000265715.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010894.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010892.hg.1 |
TC07000695.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010892.hg.1 |
ENST00000265715.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010892.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161917.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161917.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161917.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161917.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161917.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161917.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161917.hg.1 |
TC0700008760.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000440056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000460748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000477350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000480841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000492030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
ENST00000497446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008760.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161919.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161919.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161919.hg.1 |
ENST00000265715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161919.hg.1 |
NM_000441.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161919.hg.1 |
TC0700008760.hg.1 |
Clariom D TC |